Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R4/SSTR4 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB036644 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036644 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036644 Supplier Novus Biologicals Supplier No. NBP289053

Rabbit Polyclonal Antibody

Somatostatin R4/SSTR4 Polyclonal specifically detects Somatostatin R4/SSTR4 in Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Somatostatin R4/SSTR4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized
Gene Alias G-protein coupled receptor, somatostatin receptor 4, somatostatin receptor type 4, SS4R, SS-4-R, SS4-R
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 182-194 of rat Somatostatin R4/SSTR4.
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6754
Reconstitution Reconstitute with 200 μL distilled water.
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.