Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R5/SSTR5 Antibody (SR1723), Novus Biologicals™
SDP

Catalog No. NB259022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259022 100 μg
NB260312 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259022 Supplier Novus Biologicals Supplier No. NBP322300100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Somatostatin R5/SSTR5 Monoclonal antibody specifically detects Somatostatin R5/SSTR5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen Somatostatin R5/SSTR5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SR1723
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:100
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SS5R, SS-5-R, SS5-R
Host Species Rabbit
Immunogen A synthesized peptide derived from human Somatostatin R5/SSTR5 (Uniprot #: P35346)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6755
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.