Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin R5/SSTR5 Antibody (SR1723), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
| Antigen | Somatostatin R5/SSTR5 |
|---|---|
| Clone | SR1723 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
Description
Somatostatin R5/SSTR5 Monoclonal antibody specifically detects Somatostatin R5/SSTR5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
| Somatostatin R5/SSTR5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SS5R, SS-5-R, SS5-R | |
| A synthesized peptide derived from human Somatostatin R5/SSTR5 (Uniprot #: P35346) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1723 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6755 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title