Learn More
SOMNI Scientific™ SOMNI Scientific™ O2 Hose Assembly DISS
Provided with standard medical DISS Female threaded fittings on both ends The DISS fitting mates with the DISS Male on the anesthesia system
$97.50 - $188.30
Catalog Number | Mfr. No. | Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Price | Quantity & Availability | |||||
04-777-218
![]() |
SOMNI Scientific™
HF10009 |
3 ft. |
Each for $97.50
|
|
|||||
04-777-219
![]() |
SOMNI Scientific™
HF10010 |
6 ft. |
Each for $140.70
|
|
|||||
04-777-220
![]() |
SOMNI Scientific™
HF10011 |
9 ft. |
Each for $158.50
|
|
|||||
04-777-221
![]() |
SOMNI Scientific™
HF10012 |
12 ft. |
Each for $188.30
|
|
|||||
Description
- The green oxygen hose assembly is provided with standard medical DISS Female threaded fittings on both ends
- The DISS fitting mates with the DISS Male on the anesthesia system
- Medical grade DISS Oxygen fittings
Specifications
Research Animal Inhalant Anesthesia |