Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonic Hedgehog/Shh Antibody (05), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB353259 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353259 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353259 Supplier Novus Biologicals Supplier No. NBP306169APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sonic Hedgehog/Shh Monoclonal antibody specifically detects Sonic Hedgehog/Shh in Human samples. It is validated for ELISA

Specifications

Antigen Sonic Hedgehog/Shh
Applications ELISA
Classification Monoclonal
Clone 05
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Sonic Hedgehog/Shh (Uniprot#: Q15465; Met1-Gly197).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Neuroscience, Sensory Systems, Stem Cell Signaling Pathway, Stem Cells, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6469
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.