Learn More
Description
Specifications
Specifications
| Antigen | Sonic Hedgehog/Shh |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5H4 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 32867229), Immunohistochemistry-Paraffin 1:200-1:1000 |
| Formulation | Ascites with 0.03% Sodium Azide |
| Gene Accession No. | Q15465 |
| Gene Alias | HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS |
| Gene Symbols | SHH |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
