Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonic Hedgehog/Shh Antibody (5H4), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 4 publications

$206.50 - $499.50

Specifications

Antigen Sonic Hedgehog/Shh
Clone 5H4
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 32867229), Immunohistochemistry-Paraffin 1:200-1:1000
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP222126SS
SDP
View Documents
Novus Biologicals
NBP222126SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP222126
SDP
View Documents
Novus Biologicals
NBP222126
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Sonic Hedgehog/Shh
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 32867229), Immunohistochemistry-Paraffin 1:200-1:1000
Monoclonal
Ascites
RUO
Human, Mouse, Primate
Q15465
6469
Purified recombinant fragment of human Sonic Hedgehog (amino acids 26-170) expressed in E. coli. [UniProt# Q15465]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
5H4
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Neuroscience, Sensory Systems, Stem Cell Signaling Pathway, Stem Cells, Vision
Ascites with 0.03% Sodium Azide
HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
SHH
IgG1
Unpurified
49.6 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.