Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Monostearate, NF, 68-76%, Spectrum™ Chemical
SDP

Catalog No. 18606937 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
90 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-937 12 kg Poly Pail
18-606-936 2.5 kg Poly Pail
18-606-938 90 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-937 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SPA6312KG
Add to Cart
Add to Cart

SPA63, 1338-41-6, C24H46O6

Spectrum™ Chemical Sorbitan Monostearate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Ignition Residue 0.005
Packaging Poly Pail
Quantity 12 kg
Saponification Value 147-157
Percent Purity 68 to 76%
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Sorbitan Monostearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.