Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol Dehydrogenase Antibody (OTI9E7), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB274288APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274288APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274288APC Supplier Novus Biologicals Supplier No. NBP274288APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sorbitol Dehydrogenase Monoclonal specifically detects Sorbitol Dehydrogenase in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Sorbitol Dehydrogenase
Applications Flow Cytometry
Classification Monoclonal
Clone OTI9E7
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1
Gene Symbols SORD
Host Species Mouse
Immunogen Full length recombinant protein of human Sorbitol Dehydrogenase (NP_003095) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6652.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.