Learn More
Description
Specifications
Specifications
| Antigen | SorLA |
| Applications | Immunocytochemistry, Immunofluorescence, Peptide ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 10 ug/ml, Peptide ELISA Detection limit 1:32000 |
| Formulation | Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide |
| Gene Accession No. | NP_003096.1 |
| Gene Alias | C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats |
| Gene Symbols | SORL1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
