Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sortilin Antibody (334703), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB363181 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363181 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363181 Supplier Novus Biologicals Supplier No. FAB31541I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sortilin Monoclonal specifically detects Sortilin in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Sortilin
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 334703
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 100 kDa NT receptor, Glycoprotein 95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, sortilin, sortilin 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Sortilin, Ser78-Asn755, Accession # Q99523
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 6272
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.