Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOS1 Antibody (2B2), Novus Biologicals™
SDP

Catalog No. NB298362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298362 50 μL
NB298363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298362 Supplier Novus Biologicals Supplier No. NBP32630750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOS1 Monoclonal antibody specifically detects SOS1 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen SOS1
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 2B2
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SOS1 [UniProt Q07889]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6654
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.