Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOS1 Antibody (2B2), Novus Biologicals™

Rabbit Monoclonal Antibody
$295.74 - $479.73
Specifications
| Antigen | SOS1 |
|---|---|
| Clone | 2B2 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
SOS1 Monoclonal antibody specifically detects SOS1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| SOS1 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1 | |
| A synthesized peptide derived from Human SOS1 [UniProt Q07889] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2B2 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 6654 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title