Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOS1 Antibody (SOS-01) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11068800 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11068800 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11068800 Supplier Novus Biologicals Supplier No. NB11068800
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOS1 Monoclonal specifically detects SOS1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SOS1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone SOS-01
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Accession No. Q07889
Gene Alias GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1
Gene Symbols SOS1
Host Species Mouse
Immunogen Peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human Sos protein (NM_005633.3)
Molecular Weight of Antigen 150 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6654
Test Specificity The SOS-01 reacts with human Sos, an ubiquitously expressed 150 kDa intracellular protein.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.