Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOST/Sclerostin Antibody (2), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB307011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307011 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307011 Supplier Novus Biologicals Supplier No. NBP289575MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOST/Sclerostin Monoclonal antibody specifically detects SOST/Sclerostin in Human samples. It is validated for ELISA

Specifications

Antigen SOST/Sclerostin
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias sclerostin, VBCHsclerosteosis
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SOST/Sclerostin (Accession#: NP_079513.1; Gln 24-Tyr 213).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50964
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.