Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOST/Sclerostin Antibody (RM0129-4D77), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBPF06AF405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBPF06AF405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBPF06AF405 Supplier Novus Biologicals Supplier No. NBP122506AF405
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SOST/Sclerostin Monoclonal specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen SOST/Sclerostin
Applications Western Blot
Classification Monoclonal
Clone RM0129-4D77
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q99P68
Gene Alias sclerostin, VBCHsclerosteosis
Gene Symbols SOST
Host Species Rat
Immunogen Mouse recombinant protein of Sclerostin
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50964
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.