Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Clonotyping Systems
Shop All Southern Biotechnology Associates Products

Click to view available options
Conjugate:
AP
HRP
Description
Includes
Enzyme-labeled, affinity-purified goat antibodies to mouse Ig (polyvalent) for initial screening; Antibodies to mouse IgM, IgG1, IgG2a, IgG2b, IgG3, kappa, and lambda for characterizing clones that test positive; Unlabeled goat anti-mouse Ig capture antibody; Substrate (Mfr. No. 5300-04: p-nitrophenyl phosphate; Mfr. No. 5300-05: 2,2-azino-di-[3-ethylbenzthiazoline sulfonic acid] [ABTS]); Substrates also available separately
- Ideal for screening and characterizing murine hybridomas
- Use either System with Mouse Immunoglobulin Isotype Panel (Mfr. No. 5300-01) for quantifying immunoglobulin levels in hybridoma supernatants and mouse serum
Specifications
Specifications
| Quantity | 1 Kit, >1000 Tests |
| Conjugate | HRP |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | For system with Mouse immunoglobulin isotype panel |
| Content And Storage | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction