Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB005222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005222 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005222 Supplier Novus Biologicals Supplier No. NBP224486AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot.

Specifications

Antigen SOX1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1
Gene Symbols SOX1
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human SOX1 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Target Species Human, Mouse, Equine, Opossum, Primate, Xenopus species
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.