Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224466
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Canine samples. It is validated for Western Blot.Specifications
SOX1 | |
Polyclonal | |
Unconjugated | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen is 100% homologous in human, dog, chimpanzee and mouse. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/mL | |
Western Blot 0.5-3 ug/ml | |
Q80XF2 | |
SOX1 | |
A portion of amino acids 350-491 of human SOX1 was used as the immunogen. | |
0.1 mg | |
Stem Cells | |
6656 | |
Human, Mouse, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction