Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $426.00
Specifications
| Antigen | SOX1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224466SS
![]() |
Novus Biologicals
NBP224466SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224466
![]() |
Novus Biologicals
NBP224466 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Canine samples. It is validated for Western Blot.Specifications
| SOX1 | |
| Western Blot 0.5-3 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Stem Cells | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
| SOX1 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, dog, chimpanzee and mouse. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Canine | |
| Q80XF2 | |
| 6656 | |
| A portion of amino acids 350-491 of human SOX1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title