Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB337459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337459 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337459 Supplier Novus Biologicals Supplier No. NBP224486PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX1 Polyclonal antibody specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot

Specifications

Antigen SOX1
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1
Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human SOX1 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6656
Target Species Human, Mouse, Equine, Opossum, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.