Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX10 Antibody (2447A), PE, Novus Biologicals™
SDP

Catalog No. NB363906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363906 Supplier Novus Biologicals Supplier No. FAB28642P
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOX10 Monoclonal antibody specifically detects SOX10 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen SOX10
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 2447A
Conjugate PE
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
Host Species Rabbit
Immunogen E. coli-derived recombinant human SOX10, Met1-Ala118, Accession # P56693
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6663
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.