Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX10 Monoclonal Antibody (20B7), eBioscience™

Catalog No. 14592380
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-592-380 25 μg
50-112-2701 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-592-380 Supplier Invitrogen™ Supplier No. 14592380
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody 20B7 recognizes Sox10, is a member of the Sox superfamily (SRY-related HMG-box) that contain a DNA binding domain (high-mobility group domain) and functions as a transcription factor. Initially, Sox10 expression is localized to the developing neural crest and peripheral nervous system, but later is expressed in glial progenitors and mature oligodendrocytes. Sox10 is involved in stem cell maintenance, melanocyte differentiation, as well as central and peripheral gliogenesis and neurogenesis. Sox10 regulation of downstream proteins such as MASH1, PHOX2B, PDGF-alpha, and MITF is dependent on expression level. Sox10 is also expressed in multipotent vascular stem cells, melanomas, and gliomas. The 20B7 antibody is reported to recognize human and rat Sox 10, and based on homology should recognize mouse Sox10. Applications Reported: This 20B7 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry. Applications Tested: This 20B7 antibody has been tested by immunocytochemistry of formaldehyde fixed and permeabilized rat cells. This 20B7 antibody has also been tested by immunohistochemistry of human formalin-fixed paraffin embedded tissue sections using low pH antigen retreival and can be used at less than or equal to 10 μg/mL for both applications. It is recommended that the antibody be c...

SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX10
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 20B7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene SOX10
Gene Accession No. O55170, P56693, Q04888
Gene Alias cls; cls/sox10; colorless; colourless; colourless/sox10; DOM; dominant megacolon; dominant megacolon, mouse, human homolog of; golas; gos; HMG-box transcription factor Sox10; MGC15649; PCWH; Protein SOX-21; SOX10; Sox-10; sox10 protein; sox10b; Sox21; SOX9; SRY; SRY (sex determining region Y)-box 10; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY box 10; SRY-box 10; SRY-box containing gene 10; SRY-box containing protein 10; SRY-box transcription factor 10; SRY-related HMG-box gene 10; transcription factor SOX-10; Transcription factor SOX-M; WS2E; WS4; WS4C; zgc:100757
Gene Symbols SOX10
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20665, 29361, 6663
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.