Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX10, Rabbit anti-Human,Mouse, Clone: SOX10/2311R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP279770100UG
Description
SOX10 Monoclonal specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| SOX10 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P56693, P56693 | |
| SOX10 | |
| A recombinant fragment (around aa115-269) of human SOX10 protein (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| SOX10/2311R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 6663 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction