Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX10, Rabbit anti-Human,Mouse, Clone: SOX10/2311R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$316.50 - $572.50

Specifications

Antigen SOX10
Clone SOX10/2311R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010558
SDP
View Documents
Novus Biologicals
NBP27977020UG
20 μg
Each for $316.50
Only null left
Add to Cart
 
NB010557
SDP
View Documents
Novus Biologicals
NBP279770100UG
100 μg
Each for $572.50
Only null left
Add to Cart
 
Description

Description

SOX10 Monoclonal specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

SOX10
0.2 mg/mL
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Rabbit
Human, Mouse
P56693, P56693
6663
A recombinant fragment (around aa115-269) of human SOX10 protein (exact sequence is proprietary)
Primary
Store at 4°C.
SOX10/2311R
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
SOX10
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.