Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX10, Rabbit anti-Human,Mouse, Clone: SOX10/2311R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP27977020UG
Description
SOX10 Monoclonal specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
SOX10 | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
P56693, P56693 | |
SOX10 | |
A recombinant fragment (around aa115-269) of human SOX10 protein (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
SOX10/2311R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
Rabbit | |
Protein A or G purified | |
RUO | |
6663 | |
Store at 4°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction