Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SOX10 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6295
![]() ![]() |
Proteintech Group Inc
104221AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. SOX10 plays a central role in developing and maturing of glia. Specifically, SOX10 activates expression of myelin genes (such as DUSPP15 and MYRF) during oligodendrocyte (OL) maturation; thus, playing a central role in oligodendrocyte maturation and CNS myelination. Once induced MYRF cooperates with SOX10 to implement the myelination program. SOX10 is also a transcriptional activator of MITF acting synergistically with PAX3. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.Specifications
SOX10 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O55170, P56693, Q04888 | |
20665, 29361, 6663 | |
SOX10 Fusion Protein Ag0673 | |
Primary | |
-20°C | |
SOX10 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DOM, SOX10, Transcription factor SOX 10, WS2E, WS4 | |
SOX10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title