Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX11 Antibody (SOX11/3235R), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB391870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391870 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391870 Supplier Novus Biologicals Supplier No. NBP308594AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOX11 Monoclonal antibody specifically detects SOX11 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SOX11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SOX11/3235R
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11
Host Species Rabbit
Immunogen Recombinant fragment (around aa300-400) of human SOX11 protein (Uniprot: P35716 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6664
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.