Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX15 Antibody, Novus Biologicals™
SDP

Catalog No. NB10093439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093439 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093439 Supplier Novus Biologicals Supplier No. NB10093439
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SOX15 Polyclonal specifically detects SOX15 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen SOX15
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_008873.1
Gene Alias protein SOX-15, SOX20Protein SOX-20, SOX26SOX12, SOX27Protein SOX-12, SRY (sex determining region Y)-box 15, SRY (sex determining region Y)-box 20
Gene Symbols SOX15
Host Species Goat
Immunogen Peptide with sequence CSLPQSDPRLQGE corresponding to internal region according to NP_008873.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6665
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.