Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Antibody (614013), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB367090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367090 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367090 Supplier Novus Biologicals Supplier No. FAB19241S

Mouse Monoclonal Antibody

SOX17 Monoclonal antibody specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen SOX17
Applications Immunocytochemistry
Classification Monoclonal
Clone 614013
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Host Species Mouse
Immunogen E. coli-derived recombinant human SOX17, Asp177-Val414, Accession # Q9H6I2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64321
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.