Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224644SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224644SS 0.025 mg
NBP224644 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224644SS Supplier Novus Biologicals Supplier No. NBP224644SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX17 Polyclonal specifically detects SOX17 in Human, Mouse, Porcine, Chicken, Opossum, Primate samples. It is validated for Western Blot.

Specifications

Antigen SOX17
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS, 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9H6I2
Gene Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Gene Symbols SOX17
Host Species Rabbit
Immunogen A portion of amino acids 120-170 of human SOX17 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64321
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human,88%homologous inmonkey, and 62% homologous in mouse.
Target Species Human, Mouse, Pig, Chicken, Opossum, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.