Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX17 Antibody (OTI2B6), FITC, Novus Biologicals™
SDP

Catalog No. NBP274299F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274299F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274299F Supplier Novus Biologicals Supplier No. NBP274299F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX17 Monoclonal specifically detects SOX17 in Human, Mouse, Rat, Canine samples. It is validated for Immunohistochemistry.

Specifications

Antigen SOX17
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2B6
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3
Gene Symbols SOX17
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 177-414 of human SOX17.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64321.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.