Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX18 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2412890.025MG
Description
SOX18 Polyclonal specifically detects SOX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
SOX18 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SOX18 | |
Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG | |
Affinity Purified | |
RUO | |
54345 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA | |
HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
Rabbit | |
44 kDa | |
0.025 mg | |
Primary | |
SOX18 is predicted to not cross-react with other SOX proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction