Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX18 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | SOX18 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SOX18 Polyclonal specifically detects SOX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
SOX18 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
54345 | |
Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG | |
Primary | |
SOX18 is predicted to not cross-react with other SOX proteins. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
SOX18 | |
IgG | |
Affinity Purified | |
44 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title