Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX18 Antibody, Novus Biologicals™
SDP

Catalog No. NBP243708 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243708 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243708 Supplier Novus Biologicals Supplier No. NBP243708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects SOX18 in Human samples, and it is validated for Western Blotting

SOX18 Polyclonal specifically detects SOX18 in Human samples. It is validated for Western Blot.

Specifications

Antigen SOX18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
Gene Alias HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18
Gene Symbols SOX18
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SOX18. The exact sequence is proprietary.
Molecular Weight of Antigen 41 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54345
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.