Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110414 Supplier Invitrogen™ Supplier No. PA5110414
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MQRSPPGYGA QDDPPARRDC AWAPGHGAAA DTRGLAAGPA ALAAPAAPAS PPSPQRSPPR SPEPGRYGLS PAGRGERQAA DESRIRRPMN AFMVWAKDER.

SOX18 a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. This protein plays a role in hair, blood vessel, and lymphatic vessel development. Mutations in this gene have been associated with recessive and dominant forms of hypotrichosis-lymphedema-telangiectasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX18
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SOX18
Gene Accession No. P43680
Gene Alias AI385749; HLTRS; HLTS; Ra; ragged; Ragl; SOX18; Sox-18; SRY (sex determining region Y)-box 18; SRY box 18; SRY-box 18; SRY-box containing gene 18; SRY-box transcription factor 18; Sry-related HMG-box gene 18; transcription factor SOX-18
Gene Symbols SOX18
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SOX18 (NP_060889.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20672, 311723
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.