Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Antibody (245610R), Biotin, Novus Biologicals™
SDP

Catalog No. NB120391 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120391 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120391 Supplier Novus Biologicals Supplier No. FAB2018RB
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX2 Monoclonal specifically detects SOX2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.

Specifications

Antigen SOX2
Applications Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 245610R
Conjugate Biotin
Dilution Western Blot, Intracellular Staining by Flow Cytometry
Formulation PBS
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Host Species Mouse
Immunogen E.coli-derived recombinant human SOX2, Gly135-Met317, Accession # P48431
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.