Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Antibody (4G8), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP229623IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229623IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229623IR Supplier Novus Biologicals Supplier No. NBP229623IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX2 Monoclonal specifically detects SOX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SOX2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4G8
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Gene Symbols SOX2
Host Species Mouse
Immunogen Full length recombinant human SOX2 (NCBI NP_003097).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Embryonic Stem Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6657
Test Specificity Human. Based on sequence homology, this antibody is predicted to react with Mouse (96%), Chicken (93%), Sheep (99%) and Monkey/Primate species (99%). Sequence homology with other species: Zebrafish (87%), Xenopus tropicalis (87%), Xenopus laevis (88%).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.