Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Antibody (SOX2/3811R), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB380081 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380081 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380081 Supplier Novus Biologicals Supplier No. NBP308524JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SOX2 Monoclonal antibody specifically detects SOX2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen SOX2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SOX2/3811R
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Host Species Rabbit
Immunogen Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary) (Uniprot: P48431)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6657
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.