Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX2 Mouse anti-Human, Mouse, Clone: rSOX2/1792, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00

Specifications

Antigen SOX2
Clone rSOX2/1792
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123250
SDP
View Documents
Novus Biologicals
NBP30788820UG
20 μg
Each of 1 for $280.00
Only null left
Add to Cart
 
Description

Description

SOX2 Monoclonal specifically detects SOX2 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

SOX2
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
10 mM PBS with 0.05% BSA
6657
IgG1 κ
Protein A or G purified
rSOX2/1792
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse
ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.