Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX4 Antibody (SOX4/2540), HRP, Novus Biologicals™
SDP

Catalog No. NB012222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012222 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012222 Supplier Novus Biologicals Supplier No. NBP279903H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX4 Monoclonal specifically detects SOX4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SOX4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SOX4/2540
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4
Gene Symbols SOX4
Host Species Mouse
Immunogen Recombinant full-length human SOX4 protein (Uniprot: Q06945)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.