Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170446 100 μg
NB170447 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB170446 Supplier Novus Biologicals Supplier No. NBP318643100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOX4 Polyclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen SOX4
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:4000, ELISA 1:100-1:1000, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation 6 ul/IP, Immunohistochemistry-Paraffin 1:200-1:500
Formulation Whole antiserum
Gene Accession No. Q06945
Gene Alias ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human SOX4 (SRY (sex determining region Y)-box 4), using two KLH-conjugated synthetic peptides containing a sequence from the N-terminus and from the C-terminus of the protein, respectively.
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6659
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.