Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$159.60 - $400.50
Specifications
Antigen | SOX4 |
---|---|
Dilution | Western Blot 1:4000, ELISA 1:100-1:1000, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation 6 ul/IP, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SOX4 Polyclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
SOX4 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Stem Cell Markers, Stem Cells | |
Whole antiserum | |
ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
Polyclonal antibody raised in rabbit against human SOX4 (SRY (sex determining region Y)-box 4), using two KLH-conjugated synthetic peptides containing a sequence from the N-terminus and from the C-terminus of the protein, respectively. | |
Whole Antiserum |
Western Blot 1:4000, ELISA 1:100-1:1000, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation 6 ul/IP, Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q06945 | |
6659 | |
Primary | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title