Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX6 Antibody (CL5685), Novus Biologicals™
SDP

Catalog No. NBP261422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261422 100 μL
NB393109 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP261422 Supplier Novus Biologicals Supplier No. NBP261422
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX6 Monoclonal specifically detects SOX6 in Human samples. It is validated for Western Blot.

Specifications

Antigen SOX6
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL5685
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6
Gene Symbols SOX6
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids: NLDTFEHGGGHSYNHKQIE
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 55553
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.