Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX6 Antibody (CL5685), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SOX6 |
---|---|
Clone | CL5685 |
Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SOX6 Monoclonal specifically detects SOX6 in Human samples. It is validated for Western Blot.Specifications
SOX6 | |
Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
Unconjugated | |
Mouse | |
HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
SOX6 | |
IgG1 | |
Protein A purified |
CL5685 | |
Monoclonal | |
Purified | |
Human | |
55553 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: NLDTFEHGGGHSYNHKQIE | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title