Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX6 Antibody (OTI1E7), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274304V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274304V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274304V Supplier Novus Biologicals Supplier No. NBP274304V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX6 Monoclonal specifically detects SOX6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen SOX6
Applications Western Blot
Classification Monoclonal
Clone OTI1E7
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6
Gene Symbols SOX6
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-244 of human SOX6 (NP_201583) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55553.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.