Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX9 Antibody (SOX9/2398), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB387261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387261 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387261 Supplier Novus Biologicals Supplier No. NBP308399AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX9 Monoclonal antibody specifically detects SOX9 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen SOX9
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone SOX9/2398
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9
Host Species Mouse
Immunogen Recombinant human SOX9 protein fragment (around aa 393-508) (exact sequence is proprietary) (Uniprot: P48436)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6662
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.