Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SP 600125
Click to view available options
Quantity:
10 mg
Description
TB1496-RMU is an Ancillary Materials Grade version of SP 600125 (Cat. No. 1496). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Specifications
Specifications
Chemical Name or Material | Anthra[1,9-cd]pyrazol-6(2H)-one |
CAS | 129-56-6 |
Quantity | 10 mg |
Target | JNK/c-jun Inhibitors |
Molecular Formula | C14H8N2O |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction