Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-B/Surfactant Protein B Antibody, Novus Biologicals™
SDP

Catalog No. NB119905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119905 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119905 Supplier Novus Biologicals Supplier No. NBP311280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SP-B/Surfactant Protein B Polyclonal specifically detects SP-B/Surfactant Protein B in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen SP-B/Surfactant Protein B
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias 18 kDa pulmonary-surfactant protein, 6 kDa protein, PSP-B, pulmonary surfactant-associated protein B, Pulmonary surfactant-associated proteolipid SPL(Phe), SFTB3, SFTP3, SMDP1, SP-B surfactant, pulmonary-associated protein B, surfactant protein B
Host Species Rabbit
Immunogen The polyclonal antibody is raised against human-based 60-amino acid synthetic SP-B polypeptide.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6439
Target Species Human, Mouse, Pig, Bovine
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.