Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-D Antibody (1.7), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB205901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB205901 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB205901 Supplier Novus Biologicals Supplier No. NBP141050JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SP-D Monoclonal antibody specifically detects SP-D in Mouse, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen SP-D
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1.7
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Host Species Mouse
Immunogen Purified porcine surfactant protein D
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6441
Target Species Mouse, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.